Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Buffer, EDTA buffer solution pH 8,0 (0,5 mol/l) | Avantor

Buffer, EDTA buffer solution pH 8,0 (0,5 mol/l)

EDTA buffer solution pH 8,0 (0,5 mol/l), 1 KIT
Catalog # EURLEMR034001
Supplier:   EUROCLONE
Buffer, EDTA buffer solution pH 8,0 (0,5 mol/l)
Catalog # EURLEMR034001
Supplier:  EUROCLONE
CAS Number:  
Buffer, EDTA buffer solution pH 8,0 (0,5 mol/l)

EDTA buffer solution pH 8,0 (0,5 mol/l), 1 KIT

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 KIT
Product Details & Documents
Specifications

Specifications

  • Pk:
    1 KIT
  • Description:
    EDTA buffer solution pH 8,0 (0,5 mol/l)

Specifications