Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Buffer, HEPES solution (1 mol/L)

Buffer, HEPES solution (1 mol/L)

HEPES buffer solution (1 mol/l), Glass bottle, 500 mL
Catalog # EURLECM0180L
Supplier:   EUROCLONE
Buffer, HEPES solution (1 mol/L)
Catalog # EURLECM0180L
Supplier:  EUROCLONE
CAS Number:  
Buffer, HEPES solution (1 mol/L)

HEPES buffer solution (1 mol/l), Glass bottle, 500 mL

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 500 mL
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Description:
    HEPES buffer solution (1 mol/l)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.