Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Buffer, TAE solution 50X concentrate (TRIS-acetate-EDTA buffer)

Buffer, TAE solution 50X concentrate (TRIS-acetate-EDTA buffer)

TAE buffer solution 50X concentrate (TRIS-acetate-EDTA buffer), Glass bottle, 1.000 mL
Catalog # TREV9860-500-2
Marketsource
Supplier:   TREVIGEN
Buffer, TAE solution 50X concentrate (TRIS-acetate-EDTA buffer)
Catalog # TREV9860-500-2
Supplier:  TREVIGEN
Marketsource
CAS Number:  
Buffer, TAE solution 50X concentrate (TRIS-acetate-EDTA buffer)

TAE buffer solution 50X concentrate (TRIS-acetate-EDTA buffer), Glass bottle, 1.000 mL

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1.000 mL
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1.000 mL
  • Description:
    TAE buffer solution 50X concentrate (TRIS-acetate-EDTA buffer)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.