Orders LinkContactUsLinkComponent
Threaded adapters, BOLA
Threaded adapters, BOLA
Transition from GL40/S40 to GL 45
Catalog # BOHLH978-40
Supplier:   Bohlender
Threaded adapters, BOLA
Catalog # BOHLH978-40
Supplier:  Bohlender
Threaded adapters, BOLA
Threaded adapters, BOLA
Transition from GL40/S40 to GL 45
Price
SFr. 26.40
Log in for availability
1 * 1 ST
Product Details & Documents
Adaptors, made of PTFE, white, allow the use of BOLA multiple distributors for bottles with female thread GL 45 also on bottles with GL 32, GL 40 and S 40 threads.
  • Chemical resistant
  • Temperature resistant from −200 to +200 °C
Specifications
  • Description:
    Transition from GL40/S40 to GL 45
  • Seq:
    02