Orders LinkContactUsLinkComponent
Tissue forceps, Aesculap®
Tissue forceps, Aesculap®
Tissue forceps, 1×2 teeth, Straight, 115 mm
Catalog # AESCBD533R
Supplier:   Aesculap
Tissue forceps, Aesculap®
Catalog # AESCBD533R
Supplier:  Aesculap
Tissue forceps, Aesculap®
Tissue forceps, 1×2 teeth, Straight, 115 mm
Price
SFr. 26.00
Log in for availability
1 * 1 ST
Product Details & Documents
Dental tissue forceps made from stainless steel.
Specifications
  • Type:
    Straight
  • Description:
    Tissue forceps, 1×2 teeth
  • Length:
    115 mm
  • Seq:
    01