Orders LinkContactUsLinkComponent
Splinter forceps, Aesculap®
Splinter forceps, Aesculap®
Splinter and dissecting forceps, Straight, serrated, very fine tips, 115 mm
Catalog # AESCBD303R
Supplier:   Aesculap
Splinter forceps, Aesculap®
Catalog # AESCBD303R
Supplier:  Aesculap
Splinter forceps, Aesculap®
Splinter and dissecting forceps, Straight, serrated, very fine tips, 115 mm
Price
SFr. 25.70
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Type:
    Straight, serrated, very fine tips
  • Description:
    Splinter and dissecting forceps
  • Length:
    115 mm