Dissecting forceps, Aesculap®
Dissecting forceps, SEMKEN, Straight, serrated, very fine tips, 125 mm
AESCBD215R
: Aesculap
Stainless steel.
- Very good chemical resistance
- Very good temperature resistance
- Easy to clean
Delicate dissecting forceps with plateau and many types.
- Type:Straight, serrated, very fine tips
- Description:Dissecting forceps, SEMKEN
- Length:125 mm
- Seq:04