Orders LinkContactUsLinkComponent
Dressing jars, with lid
Dressing jars, with lid
50 mm, 50 mm
Catalog # BOCH8770
Supplier:   BOCHEM
Dressing jars, with lid
Catalog # BOCH8770
Supplier:  BOCHEM
Dressing jars, with lid
Dressing jars, with lid
50 mm, 50 mm
Price
SFr. 18.00
Log in for availability
1 * 1 ST
Product Details & Documents
Dressing jar either with flat or high shape, made of aluminium.
  • Very good temperature resistance
  • Good chemical resistance
  • Lightweight
Specifications
  • Ext.Ø:
    50 mm
  • Height:
    50 mm
  • Seq:
    04