Orders LinkContactUsLinkComponent
Wire baskets, square and rectangular
Wire baskets, square and rectangular
100×100×100 mm
Catalog # BOCH10020
Supplier:   BOCHEM
Wire baskets, square and rectangular
Catalog # BOCH10020
Supplier:  BOCHEM
Wire baskets, square and rectangular
Wire baskets, square and rectangular
100×100×100 mm
Price
SFr. 57.20
Log in for availability
1 * 1 ST
Product Details & Documents
These stainless steel baskets are designed for transporting, storing, cleaning and sterilising laboratory utensils.
  • Spot-welded grid 8×8×1 mm
  • Reinforced edges
  • Very good chemical and temperature resistance
Specifications
  • W×D×H:
    100×100×100 mm