Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Beakers, low form

Beakers, low form

500 ml, 110 mm, 80 mm
Catalog # BOCH8512
Supplier:   BOCHEM
Beakers, low form
Catalog # BOCH8512
Supplier:  BOCHEM
Beakers, low form
Beakers, low form

500 ml, 110 mm, 80 mm

Price
SFr. 147.00
Log in for availability
1 * 1 ST
Product Details & Documents

Stainless steel, 18/10.

  • Good chemical resistance
  • Excellent resistance to heat
  • Smooth surfaces for fast, easy cleaning

Ideal for corrosive materials. Robust and durable.

Specifications
  • Ext.Ø:
    80 mm
  • Capacity:
    500 ml
  • Height:
    110 mm