Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Beakers, low form

Beakers, low form

3000 ml, 180 mm, 150 mm
Codice catalogo BOCH8535
Nome del fornitore:   BOCHEM
Beakers, low form
Codice catalogo BOCH8535
Nome del fornitore:  BOCHEM
Beakers, low form
Beakers, low form

3000 ml, 180 mm, 150 mm

Price
SFr. 341.00
Log in per disponibilità
1 * 1 ST
Product Details & Documents

Stainless steel, 18/10.

  • Good chemical resistance
  • Excellent resistance to heat
  • Smooth surfaces for fast, easy cleaning

Ideal for corrosive materials. Robust and durable.

Specifiche
  • Ext.Ø:
    150 mm
  • Capacity:
    3000 ml
  • Height:
    180 mm