Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Beakers, low form

250 ml, 85 mm, 65 mm
Catalog # BOCH8521
Supplier:   BOCHEM
Beakers, low form
Catalog # BOCH8521
Supplier:  BOCHEM
Beakers, low form
Beakers, low form

250 ml, 85 mm, 65 mm

Price
SFr. 126.00
Log in for availability
1 * 1 ST
Product Details & Documents

Stainless steel, 18/10.

  • Good chemical resistance
  • Excellent resistance to heat
  • Smooth surfaces for fast, easy cleaning

Ideal for corrosive materials. Robust and durable.

Specifications
  • Ext.Ø:
    65 mm
  • Capacity:
    250 ml
  • Height:
    85 mm