Section lifters
150 mm, 50 mm, 10 mm
Catalog # 231-1287
Supplier: BOCHEM
Jump to:
Product Details & Documents
Blade made from stainless steel 18/10, handle made from wood.
- Flexible blade
Specifications
- Length:150 mm
- Blade width:10 mm
- Blade length:50 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



