Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Double spatulas, flexible

300 mm, 65 mm, 22 mm
Catalog # BOCH3645
Supplier:   BOCHEM
Double spatulas, flexible
Catalog # BOCH3645
Supplier:  BOCHEM
Double spatulas, flexible
Double spatulas, flexible

300 mm, 65 mm, 22 mm

Price
SFr. 10.70
Log in for availability
1 * 1 ST
Product Details & Documents

Stainless steel 18/10.

Specifications
  • Length:
    300 mm
  • Blade width:
    22 mm
  • Blade length:
    65 mm