Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Section lifters

Section lifters

150 mm, 50 mm, 15 mm
Catalog # 231-1288
Supplier:   BOCHEM
Section lifters
Catalog # 231-1288
Supplier:  BOCHEM
Section lifters
Section lifters

150 mm, 50 mm, 15 mm

Price
5.70 SFr.
Log in for availability
1 * 1 ST
Product Details & Documents

Blade made from stainless steel 18/10, handle made from wood.

  • Flexible blade
Specifications
  • Length:
    150 mm
  • Blade width:
    15 mm
  • Blade length:
    50 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.