Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Support clips

Support clips

Shaft, M12, for support clips, length 250 mm, O. Ø 15 mm
Catalog # BOCH581
Supplier:   BOCHEM
Support clips
Catalog # BOCH581
Supplier:  BOCHEM
Support clips
Support clips

Shaft, M12, for support clips, length 250 mm, O. Ø 15 mm

Price
SFr. 52.20
Log in for availability
1 * 1 ST
Product Details & Documents

These support clips are coated with ceramic inside.

  • For tubes and cylindrical vessels
  • With fitting

Ordering information: The shafts (rods) need to be ordered separately.

Specifications
  • Description:
    Shaft, M12, for support clips, length 250 mm, O. Ø 15 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.