Support clips
Shaft, M12, for support clips, length 150 mm, O. Ø 15 mm
Catalog # BOCH580
Supplier: BOCHEM
Jump to:
Product Details & Documents
These support clips are coated with ceramic inside.
- For tubes and cylindrical vessels
- With fitting
: The shafts (rods) need to be ordered separately.
Specifications
- Description:Shaft, M12, for support clips, length 150 mm, O. Ø 15 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



