Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Bossheads for frames, 90°

Bossheads for frames, 90°

Stainless steel 18/10, 13 mm
Catalog # BOCH5313
Supplier:   BOCHEM
Bossheads for frames, 90°
Catalog # BOCH5313
Supplier:  BOCHEM
Bossheads for frames, 90°
Bossheads for frames, 90°

Stainless steel 18/10, 13 mm

Price
SFr. 64.50
Log in for availability
1 * 1 ST
Product Details & Documents

Bossheads for frames, either made of stainless steel or malleable cast iron, chromium plated.

  • For rods up to 13 mm
  • Angle: 90°
  • With socket screw (M6)
Specifications
  • Material:
    Stainless steel 18/10
  • For rod Ø:
    13 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.