Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Retort rods, non-threaded

Retort rods, non-threaded

12 mm, 1000 mm
Catalog # 765-0094
Supplier:   BOCHEM
Retort rods, non-threaded
Catalog # 765-0094
Supplier:  BOCHEM
Retort rods, non-threaded
Retort rods, non-threaded

12 mm, 1000 mm

Price
SFr. 20.60
Log in for availability
1 * 1 ST
Product Details & Documents

Anodised aluminium.

Specifications
  • Ø:
    12 mm
  • Length:
    1000 mm
  • Seq:
    05
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.