Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Retort rods, non-threaded

Retort rods, non-threaded

12 mm, 600 mm
Catalog # BOCH5141
Supplier:   BOCHEM
Retort rods, non-threaded
Catalog # BOCH5141
Supplier:  BOCHEM
Retort rods, non-threaded
Retort rods, non-threaded

12 mm, 600 mm

Price
SFr. 16.20
Log in for availability
1 * 1 ST
Product Details & Documents

Anodised aluminium.

Specifications
  • Ø:
    12 mm
  • Length:
    600 mm
  • Seq:
    03
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.