Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Bossheads for frames, 90°

Bossheads for frames, 90°

Tempered cast iron, 13 mm
Catalog # BOCH5373
Supplier:   BOCHEM
Bossheads for frames, 90°
Catalog # BOCH5373
Supplier:  BOCHEM
Bossheads for frames, 90°
Bossheads for frames, 90°

Tempered cast iron, 13 mm

Price
SFr. 15.60
Log in for availability
1 * 1 ST
Product Details & Documents

Bossheads for frames, either made of stainless steel or malleable cast iron, chromium plated.

  • For rods up to 13 mm
  • Angle: 90°
  • With socket screw (M6)
Specifications
  • Material:
    Tempered cast iron
  • For rod Ø:
    13 mm