ContactUsLinkComponent
SONOREX LONGLIFE RK 102 CH, ULTRASONIC BATH, 35 KHZ, 230 V 50/60 HZ, ID 220X135X100 MM 3 L, OSCILLATING TANK OF 2 MM S/S AISI 316 TI TIMER 1-15 MIN AND CONTINUOUS, HEATING 30-80C (86-176F), DRAIN BALL COCK 1/4 , ULTRASONIC PEAK OUTPUT 480 W, W 1 * 1 items
Catalog # BAND3031
SONOREX LONGLIFE RK 102 CH, ULTRASONIC BATH, 35 KHZ, 230 V 50/60 HZ, ID 220X135X100 MM 3 L, OSCILLATING TANK OF 2 MM S/S AISI 316 TI TIMER 1-15 MIN AND CONTINUOUS, HEATING 30-80C (86-176F), DRAIN BALL COCK 1/4 , ULTRASONIC PEAK OUTPUT 480 W, W 1 * 1 items
Catalog # BAND3031
Frequently Bought Together