ContactUsLinkComponent
Ada-df 1,adapter for mounting iq sensors for measuring of ph/orp, conductivity and d.o. in d 700/n or dms/n flow-thru vessels 1 * 1 items
Catalog # WTWO203761
Supplier:  WTW ONLINE
Ada-df 1,adapter for mounting iq sensors for measuring of ph/orp, conductivity and d.o. in d 700/n or dms/n flow-thru vessels 1 * 1 items
Catalog # WTWO203761
Supplier:  WTW ONLINE
Frequently Bought Together