DECKELDICHTUNG FÜR SANOCLAV LA-MCS-203 1 * 1 ST
Catalog # SANC07014
Supplier: SANOCLAV WOLF
Frequently Bought Together
Vacuum seal PTFE / FKM
Supplier: Heidolph Instruments GmbH & Co.KG
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Accessories for Transferpette® electronic pipettes
Supplier: Brand
Accessories for electronic dispensers, Multipette E3/E3x
Supplier: EPPENDORF
VWR®, Accessories for Mechanical Pipettes
Supplier: VWR Collection
Accessories for PIPETBOY acu / pro
Supplier: Integra
Accessories for HandyStep® electronic
Supplier: Brand
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Accessories for pipetus® / pipetus®-standard / pipetus®-junior
Supplier: HIRSCHMANN
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Accessories for accu-jet® pro
Supplier: Brand
Solvent desorption tubes, Supelco®
Supplier: Merck
BIOTECTOR ACID REAGENT WITH CATALYST 1 * 20 L
Supplier: Hach
Schutzfilter 5 ml für Eppendorf Research Plus 1 * 50 ST
Supplier: EPPENDORF