Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Cyclohexanone ≥99%, GPR RECTAPUR®

Metal drum, 25 L
Catalog # 23235.462
Flammable
Irritant
   Safer Human and Environmental Health
Supplier:   VWR Chemicals
Cyclohexanone ≥99%, GPR RECTAPUR®
Catalog # 23235.462
Supplier:  VWR Chemicals
   Safer Human and Environmental Health
CAS Number:  108-94-1
Cyclohexanone ≥99%, GPR RECTAPUR®
Cyclohexanone ≥99%, GPR RECTAPUR®

Metal drum, 25 L

CAS Number:  108-94-1
Price
SFr. 818.00
Log in for availability
1 * 25 L
Product Details & Documents
Synonyms:  Pimelic Ketone
  • CAS Number:108-94-1
  • Formula:C₆H₁₀(=O)
  • MDL Number:MFCD00001625
  • Molecular Weight:98.14 g/mol
  • Merck Index:13,02755
  • Boiling Pt:155.6 °C (1013 hPa)
  • Melting Pt:–31 °C
  • Density:0.9478 g/cm³ (20 °C)
  • Flash Pt:43 °C
  • Storage Temperature:Ambient
  • EINECS:203-631-1
  • UN:1915
  • ADR:3,III
  • REACH:01-2119453616-35
Specifications

Specification Test Results

  • Assay:
    Min. 99 %
  • IR Spectrum:
    Passes test
  • Refractive index (20°C):
    1.449 - 1.451
  • Water:
    Max. 0.2 %

Specifications

  • Pack type:
    Metal drum
  • Pk:
    25 L
  • Cat. No.:
    23235.462
  • Environmental certification:
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion
  • Environmentally Preferable:
    Safer Human and Environmental Health

Specifications