Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Cyclohexanone ≥99%, GPR RECTAPUR®

Cyclohexanone ≥99%, GPR RECTAPUR®

Plastic bottle, 5 L
Catalog # 23235.360
Flammable
Irritant
   Safer Human and Environmental Health
Supplier:   VWR Chemicals
Cyclohexanone ≥99%, GPR RECTAPUR®
Catalog # 23235.360
Supplier:  VWR Chemicals
   Safer Human and Environmental Health
CAS Number:  108-94-1
Cyclohexanone ≥99%, GPR RECTAPUR®
Cyclohexanone ≥99%, GPR RECTAPUR®

Plastic bottle, 5 L

CAS Number:  108-94-1
Price
SFr. 339.00
Log in for availability
1 * 5 L
Product Details & Documents
Synonyms:  Pimelic Ketone
  • CAS Number:108-94-1
  • Formula:C₆H₁₀(=O)
  • MDL Number:MFCD00001625
  • Molecular Weight:98.14 g/mol
  • Merck Index:13,02755
  • Boiling Pt:155.6 °C (1013 hPa)
  • Melting Pt:–31 °C
  • Density:0.9478 g/cm³ (20 °C)
  • Flash Pt:43 °C
  • Storage Temperature:Ambient
  • EINECS:203-631-1
  • UN:1915
  • ADR:3,III
  • REACH:01-2119453616-35
Specifications

Specification Test Results

  • Assay:
    Min. 99 %
  • IR Spectrum:
    Passes test
  • Refractive index (20°C):
    1.449 - 1.451
  • Water:
    Max. 0.2 %

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 L
  • Cat. No.:
    23235.360
  • Environmental certification:
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion
  • Environmentally Preferable:
    Safer Human and Environmental Health

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.