Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Nitric acid 68%, TECHNICAL | Avantor

Nitric acid 68%, TECHNICAL

Plastic bottle, 5 L
Catalog # 20406.361
Oxidizing
Corrosive
Toxic
Supplier:   VWR Chemicals
Nitric acid 68%, TECHNICAL
Catalog # 20406.361
Supplier:  VWR Chemicals
CAS Number:  7697-37-2
Nitric acid 68%, TECHNICAL
Nitric acid 68%, TECHNICAL

Plastic bottle, 5 L

CAS Number:  7697-37-2
Price
SFr. 130.00
Log in for availability
1 * 5 L
Product Details & Documents
Synonyms:  Nitric acid solution
  • CAS Number:7697-37-2
  • Formula:HNO₃
  • MDL Number:MFCD00011349
  • Molecular Weight:63.01 g/mol
  • Merck Index:13,06608
  • Boiling Pt:120.5 °C (1013 hPa)
  • Melting Pt:–42 °C
  • Density:1.4 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-714-2
  • ADR:8 (5.1),II
  •  :Density: 1.51 g/cm³ (20 °C)
  • UN:2031
Specifications

Specification Test Results

  • Assay:
    68 - 70 %

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.