Hydrochloric acid 35%, TECHNICAL
Plastic bottle, 5 L
Supplier: VWR Chemicals
Jump to:
Product Details & Documents
Synonyms: Hydrochloric acid solution, Muriatic acid
- CAS Number:7647-01-0
- Formula:HCl
- MDL Number:MFCD00011324
- Molecular Weight:36.46 g/mol
- Boiling Pt:110 °C (1013 hPa)
- Melting Pt:–30 °C
- Density:1.18 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-595-7
- UN:1789
- ADR:8,II
- REACH:01-2119484862-27
Dokumente
Certificates
Search by Lot Number1L Plastic bottle for liquids DIN 45 UN X (ZPACK13961000) 2,5L Plastic bottle for liquids DIN 45 UN X (ZPACK13962500) 25L Plastic drum for liquids DIN 61 (ZPACK13952503) 5L Plastic bottle for liquids DIN 45 UN X (ZPACK13951035)
Verpackungsinformationen
P20_VWRC20246.298_SPEC_EN.pdf P20_VWRC20246.323_SPEC_EN.pdf P20_VWRC20246.367_SPEC_EN.pdf P20_VWRC20246.460_SPEC_EN.pdf spec_VWRC20246.298
Produktspezifikation
More
sds_VWRC20246
Sicherheitsdatenblatt (SDS)
VWR Chemicals Big Packs and Bulks
Produktliteratur
Specifications
Specification Test Results
- Assay:Min. 35 %
Specifications
- Pack type:Plastic bottle
- Pk:5 L



