Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Casting dams, Owl™

Casting dam, 23 mm (L), DAM-23
Catalog # OWLADAM-23
Casting dams, Owl™
Catalog # OWLADAM-23
Casting dams, Owl™
Casting dams, Owl™

Casting dam, 23 mm (L), DAM-23

Price
SFr. 197.00
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Model:
    DAM-23
  • Description:
    Casting dam, 23 mm (L)
  • Seq:
    03
  • Cat. No.:
    OWLADAM-23
  • Gel size W×L:
    230×230 mm