Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Casting dams, Owl™

Casting dams, Owl™

Casting dam, 20 mm (L), DAM-20
Catalog # OWLADAM-20
Casting dams, Owl™
Catalog # OWLADAM-20
Casting dams, Owl™
Casting dams, Owl™

Casting dam, 20 mm (L), DAM-20

Price
SFr. 197.00
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Model:
    DAM-20
  • Description:
    Casting dam, 20 mm (L)
  • Seq:
    02
  • Cat. No.:
    OWLADAM-20
  • Gel size W×L:
    200×200 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.