Orders LinkContactUsLinkComponent
Robotic tips for Tecan®
Robotic tips for Tecan®
904-252G (wide bore), 1000 µl, -, -, Black, 10 racks, 96 each
  736-0064
   Low Manufacturing Impact
Robotic tips for Tecan®
  736-0064
   Low Manufacturing Impact
Robotic tips for Tecan®
Robotic tips for Tecan®
904-252G (wide bore), 1000 µl, -, -, Black, 10 racks, 96 each
SFr. 186.00
1 * 960 ST
High quality tips, especially intended for research in molecular biology laboratories.
  • Transparent or black PP
  • Tips with ART® filters: Eliminate any risk of cross-contamination between samples; designed for applications with DNA, RNA as well as radioactive and infectious samples
  • Certified free of RNase, DNase, DNA and pyrogens
  • Sterile tips: Sterilised by gamma radiation
  • Wide aperture tips: Designed for applications with more viscous samples
Ordering information: Available in boxes of 10 racks or 50 racks (Econopak).
  • Packed:
    10 racks, 96 each
  • Sterile:
    -
  • Volume:
    1000 µl
  • Tip style:
    904-252G (wide bore)
  • Tip colour:
    Black
  • Filter:
    -
  • Cat. No.:
    736-0064
  • Environmental certification:
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • Environmentally Preferable:
    Low Manufacturing Impact
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.