Orders LinkContactUsLinkComponent
Wire triangles
Wire triangles
With smooth earthenware pipes, tube length 40 mm
Catalog # HECH41065004
Wire triangles
Catalog # HECH41065004
Wire triangles
Wire triangles
With smooth earthenware pipes, tube length 40 mm
Price
SFr. 7.00
Log in for availability
1 * 1 ST
Product Details & Documents
These wire triangles with plain clay tubes are made of galvanised steel wire.
Specifications
  • Description:
    With smooth earthenware pipes, tube length 40 mm
  • Seq:
    01