Storage Drawers
Single drawer, outer dimension: 164×134×74 mm, 1 drawer, 1,20 L, C, 155×120×64 mm
216-4345
Stackable polystyrene drawer unit ideal for organising and storing laboratory components, consumables, and various materials.
- Can be wall-mounted, fixed to a partition or placed on a countertop
- Single or double drawer
- Various sizes available
- Transparent drawers with label holder
- Compartmentalised drawers
The modular assembly system makes these pull-out drawers easy to open.
- Type:C
- Capacity:1,20 L
- Description:Single drawer, outer dimension: 164×134×74 mm, 1 drawer
- Seq:03
- W×D×H:155×120×64 mm



