Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Storage Drawers

Storage Drawers

Double drawer (stacked), outer dimension: 164×134×74 mm, 2 drawers, 0,50 L, B, 155×120×54 mm
Catalog # 216-4362
Supplier:   MULTIROIR
Storage Drawers
Catalog # 216-4362
Supplier:  MULTIROIR
Storage Drawers
Storage Drawers

Double drawer (stacked), outer dimension: 164×134×74 mm, 2 drawers, 0,50 L, B, 155×120×54 mm

Price
SFr. 39.80
Log in for availability
1 * 1 ST
Product Details & Documents

Stackable polystyrene drawer unit ideal for organising and storing laboratory components, consumables, and various materials.

  • Can be wall-mounted, fixed to a partition or placed on a countertop
  • Single or double drawer
  • Various sizes available
  • Transparent drawers with label holder
  • Compartmentalised drawers

The modular assembly system makes these pull-out drawers easy to open.

Specifications
  • Type:
    B
  • Capacity:
    0,50 L
  • Description:
    Double drawer (stacked), outer dimension: 164×134×74 mm, 2 drawers
  • Seq:
    02
  • W×D×H:
    155×120×54 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.