ContactUsLinkComponent
5ml graduated Pasteur pipette, 1.0ml div, 217mm, pk250 1 * 250 items
Catalog # 612-0012
Supplier:  Alpha Laboratories
5ml graduated Pasteur pipette, 1.0ml div, 217mm, pk250 1 * 250 items
Catalog # 612-0012
Supplier:  Alpha Laboratories
Frequently Bought Together