Orders LinkContactUsLinkComponent
Tripods
Tripods
With screwed feet, tempered cast iron (zincked), 100×180 mm
Catalog # BOCH5230
Supplier:   BOCHEM
Tripods
Catalog # BOCH5230
Supplier:  BOCHEM
Tripods
Tripods
With screwed feet, tempered cast iron (zincked), 100×180 mm
Price
SFr. 33.10
Log in for availability
1 * 1 ST
Product Details & Documents
Tripods made from stainless steel or tempered cast iron (zincked).
  • With welded or screwed feet
Specifications
  • Int.Ø×H:
    100×180 mm
  • Description:
    With screwed feet, tempered cast iron (zincked)