Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Sample bottles without push-on Closure

Sample bottles without push-on Closure

11,5 ml, 50 mm, 22 mm
Catalog # 215-2398
Supplier:   GRAVIS
Sample bottles without push-on Closure
Catalog # 215-2398
Supplier:  GRAVIS
Sample bottles without push-on Closure
Sample bottles without push-on Closure

11,5 ml, 50 mm, 22 mm

Price
SFr. 144.00
Log in for availability
1 * 220 ST
Product Details & Documents

Chemistry glass, white or amber, without push-on cap.

  • Amber glass to protect light-sensitive materials

Ordering information: Push-on PE caps are available separately.

Documents

Specifications
  • Capacity:
    11,5 ml
  • Height:
    50 mm
  • Neck-Ø:
    22 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.