Orders LinkContactUsLinkComponent
Measuring beaker with handle
Measuring beaker with handle
2000 ml
Catalog # NICKJUGS2000H
Supplier:   NICKEL ELECTRO
Measuring beaker with handle
Catalog # NICKJUGS2000H
Supplier:  NICKEL ELECTRO
Measuring beaker with handle
Measuring beaker with handle
2000 ml
Price
SFr. 98.40
Log in for availability
1 * 1 ST
Product Details & Documents
Stainless steel, rust-proof.
  • Very good chemical resistance
  • Very good thermal resistance
  • Smooth surfaces for fast, easy cleaning
Conical, without rim, with broad base and textured graduations.
Specifications
  • Capacity:
    2000 ml