Orders LinkContactUsLinkComponent
96-Multiwell insert system, Falcon®, Corning
96-Multiwell insert system, Falcon®, Corning
Five insert plates with feeder trays and lids
Catalog # 734-0368
   Low Manufacturing Impact
Supplier:   Corning
96-Multiwell insert system, Falcon®, Corning
Catalog # 734-0368
Supplier:  Corning
   Low Manufacturing Impact
96-Multiwell insert system, Falcon®, Corning
96-Multiwell insert system, Falcon®, Corning
Five insert plates with feeder trays and lids
Price
SFr. 2’950.00
Log in for availability
1 * 5 ST
Product Details & Documents
The Falcon® HTS 96-multiwell insert system is a cell culture insert platform suitable for both manual and robotic screening of compounds in cell-based assays. The system has been tested for its ability to produce a differentiated monolayer of Caco-2, LLC-PK1, and MDCK cells making it an ideal platform for in vitro bioavailability and permeability studies. This automation compatible platform is composed of a 1,0 µm pore size PET membrane-based 96-multiwell insert plate, a media feeder tray and a lid. The drop-in baffle for the feeder tray mitigates media sloshing and lowers the risk of contamination.
  • Automation compatible design suitable for use with most robots and fluid handling instruments
  • Excellent reproducibility - the one-piece feeder tray enhances consistency in well-to-well monolayer growth
  • Ideal for drug transport studies
Specifications
  • Description:
    Five insert plates with feeder trays and lids
  • Cat. No.:
    734-0368
  • Environmental certification:
    ISO 14001 - Bedford
  • Environmentally Preferable:
    Low Manufacturing Impact
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • 96-square well, angled-bottom plate, with lid
    96-square well, angled-bottom plate, with lid
    734-1378
    Log in for availability
    1 * 5 ST - SFr. 606.00
Product Information
  • Catalog #
    Availability
    Price
  • Feeder tray, PS, non-treated, designed for use with Falcon® 96-multiwell insert systems
    Feeder tray, PS, non-treated, designed for use with Falcon® 96-multiwell insert systems
    736-2024
    Log in for availability
    1 * 5 ST - SFr. 214.00