Orders LinkContactUsLinkComponent
Clamps
Clamps
60 mm
Catalog # 241-7190
Supplier:   BOCHEM
Clamps
Catalog # 241-7190
Supplier:  BOCHEM
Clamps
Clamps
60 mm
Price
SFr. 25.70
Log in for availability
1 * 1 ST
Product Details & Documents
Malleable cast iron, chromed.
  • Jaws have cork inserts to protect clamped items
  • Without bosshead for fixing to stand
  • Wing screws for rapid, easy clamping
DIN 12894
Certifications: DIN 12894
Specifications
  • Spanning width:
    60 mm