Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0.00 SFr.
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
Closures
Bottle Closures
Screw caps, bakelite
Print
Screw caps, bakelite
with TRISEAL seal, 40 mm, Black
Catalog # SPVAP09040
Supplier:
GRAVIS
Screw caps, bakelite
Catalog # SPVAP09040
Supplier:
GRAVIS
Return to Top
Screw caps, bakelite
with TRISEAL seal, 40 mm, Black
Specifications
Jump to:
Specifications
Price
SFr. 90.50
Log in
for availability
1 * 100 ST
-
+
Other Product Family Options Available
Return Policy
Product Details & Documents
SPVAP08028_TDS_EN
Specifications
Thread:
40 mm
Colour:
Black
Description:
with TRISEAL seal
Seq:
21
Sample bottles
#: 215-2637
Supplier: GRAVIS