Tissue processing/embedding cassettes, Histosette® II, M485
Blue
Codice catalogo 720-0230
Nome del fornitore: Simport Scientific
Tissue processing/embedding cassettes, Histosette® II, M485
Codice catalogo 720-0230
Nome del fornitore: Simport Scientific
Vai a
Product Details & Documents
Acetal polymer, similar to M492, but covers are already removed from cassettes and are packed separately in the case.
- Convenient for cassette labelling instruments as the covers are already removed
- Resistant to the chemical action of histological solvents
- Flow-through slots maximise fluid exchange
Specifiche
- Colour:Blue
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.



