Orders LinkContactUsLinkComponent
Turbidity standards, Stablcal®
Turbidity standards, Stablcal®
Stablcal® stabilised formazine standard, 20 NTU, 1 L
N° de catalogue 708-0075
Fournisseur:   Hach
Turbidity standards, Stablcal®
N° de catalogue 708-0075
Fournisseur:  Hach
Numéro CAS:  
Turbidity standards, Stablcal®
Turbidity standards, Stablcal®
Stablcal® stabilised formazine standard, 20 NTU, 1 L
Numéro CAS:  
Prix
SFr. 459.00
Connexion Pour la disponibilité
1 * 1 L
Product Details & Documents
Stablcal® turbidity standards for all turbidimeters. USEPA accepted for calibration purposes. Stablcal® stabilised formazine turbidity standards are true formazine dilutions developed for use in any turbidimeter.
  • Precise, pre-mixed formazine solutions
  • Based on formazine primary standards
  • Enhanced stability saves time and labour
  • Confidence in results
Documents
Spécifications
Spécifications
  • Pk:
    1 L
  • Turbidity:
    20 NTU
  • Description:
    Stablcal® stabilised formazine standard
Spécifications