Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Plain swabs

Plain swabs

Plain swabs, Wood/cotton, 2,5 mm, 150 mm
Catalog # 115-0502
Supplier:   BAILLY
Plain swabs
Catalog # 115-0502
Supplier:  BAILLY
Plain swabs
Plain swabs

Plain swabs, Wood/cotton, 2,5 mm, 150 mm

Price
SFr. 15.20
Log in for availability
1 * 50 ST
Product Details & Documents

Plain swabs suitable for a wide range of applications.

Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904

Specifications
  • Ø:
    2,5 mm
  • Description:
    Plain swabs
  • Length:
    150 mm
  • Seq:
    003
  • Shaft/head:
    Wood/cotton
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.