Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Plain swabs

Plain swabs, Wood/cotton, 2,2 mm, 150 mm
Catalog # 115-0500
Supplier:   BAILLY
Plain swabs
Catalog # 115-0500
Supplier:  BAILLY
Plain swabs
Plain swabs

Plain swabs, Wood/cotton, 2,2 mm, 150 mm

Price
SFr. 9.40
Log in for availability
1 * 100 ST
Product Details & Documents

Plain swabs suitable for a wide range of applications.

 : *The product(s) on this page are outside the scope of the EU Directive 2019/904

Specifications
  • Ø:
    2,2 mm
  • Description:
    Plain swabs
  • Length:
    150 mm
  • Seq:
    002
  • Shaft/head:
    Wood/cotton