Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Agar, powder, TECHNICAL

Agar, powder, TECHNICAL

Plastic bottle for solids, 1 kg
Catalog # 20768.292
Supplier:   VWR Chemicals
Agar, powder, TECHNICAL
Catalog # 20768.292
Supplier:  VWR Chemicals
CAS Number:  9002-18-0
Agar, powder, TECHNICAL
Agar, powder, TECHNICAL

Plastic bottle for solids, 1 kg

CAS Number:  9002-18-0
Price
320.00 SFr.
Log in for availability
1 * 1 kg
Product Details & Documents

Not suitable for microbiological purposes

Synonyms:  Agar Agar
  • CAS Number:9002-18-0
  • MDL Number:MFCD00081288
  • Merck Index:13,00184
  • Melting Pt:90 °C
  • Storage Temperature:Ambient
  • EINECS:232-658-1
Specifications

Specification Test Results

  • Identification:
    Passes test

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg

Specifications