Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Hydrochloric acid 35%, TECHNICAL

Plastic bottle, 1 L
Catalog # 20246.298
Corrosive
Irritant
Supplier:   VWR Chemicals
Hydrochloric acid 35%, TECHNICAL
Catalog # 20246.298
Supplier:  VWR Chemicals
CAS Number:  7647-01-0
Hydrochloric acid 35%, TECHNICAL
Hydrochloric acid 35%, TECHNICAL

Plastic bottle, 1 L

CAS Number:  7647-01-0
Price
SFr. 23.70
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Hydrochloric acid solution, Muriatic acid
  • CAS Number:7647-01-0
  • Formula:HCl
  • MDL Number:MFCD00011324
  • Molecular Weight:36.46 g/mol
  • Boiling Pt:110 °C (1013 hPa)
  • Melting Pt:–30 °C
  • Density:1.18 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-595-7
  • UN:1789
  • ADR:8,II
  • REACH:01-2119484862-27
Specifications

Specification Test Results

  • Assay:
    Min. 35 %

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.