Hydrobromic acid 47%, GPR RECTAPUR®
Glass bottle, 1 L
: Hydrogen bromide
- :10035-10-6
- :HBr
- :MFCD00011323
- :80.91 g/mol
- :1.47 g/cm³ (20 °C)
- :Ambient
- :233-113-0
- :1788
- :8,II
- Assay:Min. 47 %
- Phosphate, Phosphite (as PO4):max 0.002%
- Sulphate and sulphite (as SO4):max 0.005%
- Heavy metals (as Pb):max 0.001%
- Fe (Iron):max 0.0005%
- Pack type:Glass bottle
- Pk:1 L



