Orders LinkContactUsLinkComponent
Test kits, drop count tests | Avantor
Test kits, drop count tests
Dissolved oxygen, model OX-2P, 1 to 20 mg/l O₂, 1 KIT
Catalog # HACH146900
Supplier:   Hach
Test kits, drop count tests
Catalog # HACH146900
Supplier:  Hach
CAS Number:  
Test kits, drop count tests
Test kits, drop count tests
Dissolved oxygen, model OX-2P, 1 to 20 mg/l O₂, 1 KIT
CAS Number:  
Price
SFr. 308.00
Log in for availability
1 * 1 KIT
Product Details & Documents
The drop count test is a simple, easy and cost-effective way to test for many parameters like chlorine, chloride, dissolved oxygen, carbon dioxide, sulphite, nitrogen, formaldehyde, and more.
  • Portable - ideal for use in field or in lab
  • Used as a quick reference or for confirmatory test with colour indicator
  • Reagents supplied with the kit
Achtung! Bitte beachten: Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.
Specifications
Specifications
  • Pk:
    1 KIT
  • Description:
    Dissolved oxygen, model OX-2P
  • Range:
    1 to 20 mg/l O₂
Specifications
Empfehlungen
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen..