Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Solid phase extraction cartridges, Bond Elut™ SI | Avantor

Solid phase extraction cartridges, Bond Elut™ SI

Bond Elut™ Jr, Silica, 60 Å, 40 µm, 500 mg
  551-0375
 :   VARIAN
Solid phase extraction cartridges, Bond Elut™ SI
  551-0375
 :  VARIAN
Solid phase extraction cartridges, Bond Elut™ SI

Bond Elut™ Jr, Silica, 60 Å, 40 µm, 500 mg

 
 
   
1 * 100 ST
 

Native silica is generally regarded as the most polar SPE sorbent available. Bond Elut™ SI is particularly effective at separating compounds with a very similar structure. Applying the analytes in a non-polar solvent, then increasing the solvent polarity by increasing the concentration of a polar modifier, such as THF or ethyl acetate, delivers effective separations.

  • Highly polar phase retains polar molecules from non-polar matrices
  • High purity silica
  • Separate compounds with very similar structures
 
  • Particle Size:
    40 µm
  • Pore size:
    60 Å
  • Description:
    Bond Elut™ Jr
  • Seq:
    17
  • Bed weight:
    500 mg
  • Phase:
    Silica
  • Cat. No.:
    551-0375
  • Endcapped:
    No
  • Surface area:
    600 m²/g
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.